Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)

G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)

Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl), 1 L
Catalog # CA82021-480
Supplier:   G-Biosciences
Supplier #:  R017
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Catalog # CA82021-480
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)

Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl), 1 L

CAS Number:  
Price
$327.56
Log in for availability
Each (1l)
Minimum order quantity is 1
Product Details & Documents

Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.

  • 1 M Tris·HCl (pH 7.4)

  • 1.5 M sodium chloride

Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.

Specifications

Specifications

  • Désignation:
    Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl)
  • Taille:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.