Labels, Brady
Printer labels, F properties, Vinyl, Tamper resistant indicating, 38.1 x 25.4 mm
Catalog # CA80090-196
Supplier:   Avantor
Labels, Brady
Catalog # CA80090-196
Supplier:  Avantor
Labels, Brady
Labels, Brady
Printer labels, F properties, Vinyl, Tamper resistant indicating, 38.1 x 25.4 mm
Price
$88.94
Log in pour la disponibilité
Each
Product Details & Documents
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable. This material has a permanent acrylic adhesive and is thermal transfer printable.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-427: Self-Laminating Vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-351: Tamper-Resistant Vinyl is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
 : RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved.
Spécifications
  • Pour l'utilisation avec:
    Tamper resistant indicating
  • Désignation:
    Printer labels, F properties
  • Dimensions:
    38.1 x 25.4 mm
  • Matière:
    Vinyl
  • Cat. No.:
    CA80090-196