Labels, Brady
Portable thermal labels, Nylon Cloth, 12.7×38.1 mm (0.5×1.5"), 500 labels/roll
Catalog # CA80090-086
Supplier: Avantor
Jump to:
Product Details & Documents
Labels for the TLS 2200 and TLS-PC Link Thermal Transfer Portable Printers.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Please Note: This material is UL, CSA and AGA approved with its respective ribbon. See the 'Recommended Ribbon Series' for the appropriate ribbon to use.
Spécifications
- Désignation:Portable thermal labels
- Dimensions:12.7×38.1 mm (0.5×1.5")
- Matière:Nylon Cloth
- Packaging:500 labels/roll