Polypropylene Vials, Starplex®
Labeled, 10.00 ml, Flat bottom, Clear, Oui, Screw cap, attached, in plastic peel pack, White, Sterile
Catalog # CA73270-066
Supplier: Starplex
: V102-POUCH
Jump to:
Product Details & Documents
These vials are made of crack-resistant medical grade polypropylene.
- With or without screw caps
- Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
Spécifications
- Avec bouchon:Oui
- Cap Type:Screw cap, attached, in plastic peel pack
- Sterility:Sterile
- Capacité:10.00 ml
- Color:Clear
- Désignation:Labeled
- Cap Color:White
- Bottom Style:Flat bottom