Polypropylene Vials, Starplex®
Unlabeled, 10.00 ml, Flat bottom, Clear, Oui, Screw cap, packed separately, White, Non sterile
CA73240-127
: Starplex
: V102-NL
These vials are made of crack-resistant medical grade polypropylene.
- With or without screw caps
- Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
- Avec bouchon:Oui
- Cap Type:Screw cap, packed separately
- Sterility:Non sterile
- Capacité:10.00 ml
- Color:Clear
- Désignation:Unlabeled
- Cap Color:White
- Bottom Style:Flat bottom