Accessories for Centrifuge, Iris Sample Processing
Inserts for 4–5 mL (13 x 75 mm, 10.25 x 68 mm) Tubes
Catalog # CA56430-142
Supplier: Statspin
Supplier #: SV01
Jump to:
Product Details & Documents
Spécifications
- Désignation:Inserts for 4–5 mL (13 x 75 mm, 10.25 x 68 mm) Tubes