Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Adapter, for 1 HPLC vessel 1.5 ml, for rotor F-45-18-17-Cryo, 18 pieces
Catalog # CA5427708006
Supplier: Eppendorf
: 5427708006
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Catalog # CA5427708006
Supplier: Eppendorf
Jump to:
Product Details & Documents
Spécifications
- Désignation:Adapter, for 1 HPLC vessel 1.5 ml, for rotor F-45-18-17-Cryo, 18 pieces
- Cat. No.:CA5427708006
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Rotor key, 5 mmRotor key, 5 mmCA53513-248Centrifuges 5417 C/R, 5418/R, 5424/R, 5430/R and 5702/R/RHLog in pour la disponibilitéEach - $42.30