Accessories for Micro-Osmometer, Advanced Instruments
Accessories for Micro-Osmometer, Advanced Instruments
Clinitrol™ 290 Reference Solution, 2 mL
Catalog # CA52196-126
Supplier #:  3MA029
Accessories for Micro-Osmometer, Advanced Instruments
Catalog # CA52196-126
Accessories for Micro-Osmometer, Advanced Instruments
Clinitrol™ 290 Reference Solution, 2 mL
Price
$482.10
Log in for availability
Pack of 10 (2ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Documents
Specifications
  • Désignation:
    Clinitrol™ 290 Reference Solution, 2 mL