Measuring Spoon, Hach | Avantor
Measuring Spoon, Hach
Plastic, 1.0 g
Catalog # CA51000
Supplier:   Hach
Fournisseur #:  51000
Measuring Spoon, Hach
Catalog # CA51000
Supplier:  Hach
Measuring Spoon, Hach
Plastic, 1.0 g
Price
$14.28
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Measuring spoon features a molded plastic cup. Weight determination is based on sodium chloride.
Spécifications
  • Capacité:
    1.0 g
  • Matière:
    Plastic