Embedding Cassettes, Simport Scientific
These disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures
The cassettes fit securely in microtome chuck adapters. They are molded from a high-density acetal polymer that is totally resistant to the chemical action of histological solvents, except for clear cassette 82028-100, which is made of polypropylene. The cassettes are designed to accept standard metal lids and will keep specimens safe during processing. The 45° anterior, slanted writing surface accepts markings easily, permitting sample identification throughout all stages of embedding. The cassettes can also be used for automatic cassette labeling instruments.
: Cassettes are packed in three dispenser boxes of 500 cassettes.
- Color:Pink
- Désignation:Embedding Cassettes