Round Bottom Flasks
250 ml, 19/26, Round
Catalog # CA47800-120
Supplier #: 015890590
Jump to:
Discontinued
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
- ST Joint No.:19/26
- Capacité:250 ml
- Bottom Style:Round