Corning® Lymphocyte Separation Medium, Corning
Lymphocyte Separation Medium, 100 ml
Supplier: Corning
Supplier #: 25-072-CI
Jump to:
Product Details & Documents
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.
Specifications
- Volume:100 ml
- Désignation:Lymphocyte Separation Medium
- Cat. No.:CA45000-726
- Storage conditions:Ambient
- Shipping conditions:Ambient