Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Trypsin | Avantor

Corning® Trypsin

Trypsin/EDTA Solution, 1x 0.05% Trypsin with 0.53 mM EDTA, 500 ml
Catalog # CA45000-662
Supplier:   Corning
Fournisseur #:  25-052-CV
Corning® Trypsin
Catalog # CA45000-662
Supplier:  Corning
CAS Number:  
Corning® Trypsin
Corning® Trypsin

Trypsin/EDTA Solution, 1x 0.05% Trypsin with 0.53 mM EDTA, 500 ml

CAS Number:  
Price
$403.00
Log in pour la disponibilité
Pack of 6 (500ml)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin.

  • Storage: -25°C to -15°C

Trypsin has a pH specification range from 7.2 to 8. At this range, phenol red inclusion gives the solution a pink color. During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not affect the functionality of the trypsin. By adjusting the pH to 7.2 – 8 with NaOH the trypsin activity could be restored.

Spécifications

Spécifications

  • Désignation:
    Trypsin/EDTA Solution, 1x 0.05% Trypsin with 0.53 mM EDTA
  • Taille:
    500 ml

Spécifications