Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Minimum Essential Medium (MEM), Powder with Earle's salts, L-glutamine, and nonesstential amino acids without sodium bicarbonate
Supplier: Corning
Fournisseur #: 50-011-PB
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Catalog # CA45000-594
Supplier: Corning
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
Documents
Certificates
Recherche par numéro de lotSpécifications
Spécifications
- Désignation:Minimum Essential Medium (MEM), Powder with Earle's salts, L-glutamine, and nonesstential amino acids without sodium bicarbonate
- Cat. No.:CA45000-594
- Storage temperature:Refrigerate
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



