Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning | Avantor

Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning

Minimum Essential Medium (MEM) Alpha Medium, Powder with Earle's salts and L-glutamine without ribonucleosides, deoxyribonucleosides, and sodium bicarbonate, Powder, 50 L
Catalog # CA45000-504
Supplier:   Corning
Fournisseur #:  50-012-PC
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Catalog # CA45000-504
Supplier:  Corning
CAS Number:  
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning

Minimum Essential Medium (MEM) Alpha Medium, Powder with Earle's salts and L-glutamine without ribonucleosides, deoxyribonucleosides, and sodium bicarbonate, Powder, 50 L

CAS Number:  
Price
$121.00
Log in pour la disponibilité
Each (50l)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.

Certifications: Triple 0.1 µm sterilized.

Spécifications

Spécifications

  • Format du milieu:
    Powder
  • Désignation:
    Minimum Essential Medium (MEM) Alpha Medium, Powder with Earle's salts and L-glutamine without ribonucleosides, deoxyribonucleosides, and sodium bicarbonate
  • Taille:
    50 L
  • Cat. No.:
    CA45000-504
  • Storage temperature:
    2...8 °C

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.