Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Medium 199 (Mod.), Corning

Corning® Medium 199 (Mod.), Corning

Medium 199 (Mod.) with Earle's salts and L-glutamine, Liquide
Catalog # CA45000-378
Supplier:   Corning
Supplier #:  10-060-CV
Corning® Medium 199 (Mod.), Corning
Catalog # CA45000-378
Supplier:  Corning
CAS Number:  
Corning® Medium 199 (Mod.), Corning
Corning® Medium 199 (Mod.), Corning

Medium 199 (Mod.) with Earle's salts and L-glutamine, Liquide

CAS Number:  
Price
$189.00
Log in pour la disponibilité
Pack of 6 (500ml)
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.

Spécifications

Spécifications

  • Format du milieu:
    Liquide
  • Désignation:
    Medium 199 (Mod.) with Earle's salts and L-glutamine
  • Cat. No.:
    CA45000-378
  • Storage temperature:
    2...8 °C

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.