Sampling Tubes and Systems, Drager
Diffusion sampler ORSA 5
Catalog # CA34330-184
Supplier: Draeger
Fournisseur #: 6728891
Jump to:
Product Details & Documents
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
Spécifications
- Désignation:Diffusion sampler ORSA 5