Sampling Tubes and Systems, Drager | Avantor
Sampling Tubes and Systems, Drager
Diffusion sampler ORSA 5
Catalog # CA34330-184
Supplier:   Draeger
Fournisseur #:  6728891
Sampling Tubes and Systems, Drager
Catalog # CA34330-184
Supplier:  Draeger
Sampling Tubes and Systems, Drager
Sampling Tubes and Systems, Drager
Diffusion sampler ORSA 5
Price
$1,112.45
Log in pour la disponibilité
Each
Product Details & Documents
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
Spécifications
  • Désignation:
    Diffusion sampler ORSA 5