PIPETS GRADUATED 138MM CS500
Catalogue # CA30002-066
Fournisseur: Copan Diagnostics
PIPETS GRADUATED 138MM CS500
Catalogue # CA30002-066
Fournisseur: Copan Diagnostics
Supplier Number: 218CS01
Frequently Bought Together
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
VWR® Disposable Serological Pipettes
Fournisseur: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
VWR® Universal Pipette Tips
Fournisseur: VWR International
VWR® Nitrile Examination Gloves, Blue
Fournisseur: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Fournisseur: Corning
VWR® Disposable Petri Dishes
Fournisseur: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH