Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

VWR® Corks

38 mm, 30, 50 mm, 57 mm
  CA23420-570
VWR® Corks
  CA23420-570
VWR® Corks
VWR® Corks

38 mm, 30, 50 mm, 57 mm

 
$3,450.72
   
Pack of 100
 

XXXX Corks function as a closure for bottles, vials, laboratory and other openings.

  • Regular length taper
  • Assorted sizes

The XXXX premium grade corks have fewer lenticels (crevices) than standard grade

Selected grade of fine textured corks. Sizes 22 through 30 have cemented construction.

This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.

 
  • Hauteur:
    38 mm
  • Stopper No.:
    30
  • Top Diameter:
    57 mm
  • Bottom Diameter:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.