Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Olympic™ Stacking Bins, Metro™ | Avantor

Olympic™ Stacking Bins, Metro™

Bentron bin, Black, 10.5W x 18.8L x 7.6H cm (41/8 x 73/8 x 3")
Catalog # CA22221-320
Supplier:   Metropolitan Wire
Fournisseur #:  SB90743CAS
Olympic™ Stacking Bins, Metro™
Catalog # CA22221-320
Supplier:  Metropolitan Wire
Olympic™ Stacking Bins, Metro™
Olympic™ Stacking Bins, Metro™

Bentron bin, Black, 10.5W x 18.8L x 7.6H cm (41/8 x 73/8 x 3")

Price
$27.64
Log in pour la disponibilité
Each
Product Details & Documents

Bins are durable, stackable, and feature high impact strength

  • A flat surface on the front of the bin is provided for easy placement of labels and bar codes
  • All bins have an open hopper for easy access to contents
  • Bins feature a lip on the edge of the rear panel for hanging

Choose from Benstat™ or Bentron™ bins:
Benstat is a static-dissipative polypropylene-based material with a surface resistivity of >105ohms/sq. and <1012ohms/sq. All Benstat bins are blue.
Bentron is a polypropylene-based material with a carbon additive to provide an electrically conductive path through the material. Bentron has a surface resistivity of <105ohms/sq. All Bentron bins are black.

Spécifications
  • Color:
    Black
  • Désignation:
    Bentron bin
  • Dimensions:
    10.5W x 18.8L x 7.6H cm (41/8 x 73/8 x 3")
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.