Super Erecta® Stem Casters, Metro™
Product Details & Documents
For use with Super Erecta® shelving and SiteSelect® posts to create mobile units. Swivel, foot-operated brake, and rigid styles are available.
- Resilient rubber tread casters are non-marking and feature a molded, soft tread that provides quiet operation
- Polyurethane tread casters are abrasion-resistant, non-marking, and shock-absorbant
Casters with high modulus rubber tread provide extra shock protection and have excellent resistance to oil and grease. Special cart-washable casters are constructed of stainless steel components. Casters include grease seals and zerk fittings to withstand high-pressure washings.
Conductive casters conform to Federal Specification FF-C-88C regarding the acceptable standards for conductive wheels. To ensure proper conductivity, aluminum split sleeves must be used with each shelf.
: Rigid casters are held in position by a connecting channel; when ordering rigid casters, shelf width must be provided. An 8.9 cm (3¹/₂") diameter donut is furnished standard on each caster.
Specifications
- Largeur:1"
- Désignation:Chrome-Plated Mobile Post
- Hauteur:63"
- Shipping Weight:3.5 lbs.
- Weight:1.6 kg (3.5 lbs.)



