Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes | Avantor

Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes

Blue, 30000×g, 500 tubes (1 bag×500 tubes)
Catalog # CA21008-953
Supplier:   Eppendorf
Fournisseur #:  022363247
Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes
Catalog # CA21008-953
Supplier:  Eppendorf
Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes
Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes

Blue, 30000×g, 500 tubes (1 bag×500 tubes)

Price
$137.00
Log in pour la disponibilité
Pack of 500
Product Details & Documents

Eppendorf® Safe-Lock 1.5 ml microtubes ensure secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1.5 ml capacity.

  • Hinged Safe-Lock lid prevents inadvertent lid opening during incubation and storage
  • g-Safe® technology, depending on the Safe-Lock Tube variant (refer to the manual), ensures outstanding centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss due to tube breakage, and provides additional safety when handling hazardous samples
  • Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures
  • Precision lid sealing minimizes evaporation rates during extended storage periods
  • Generously sized frosted lid and side surface facilitate easy labeling
  • Assured functionality in a temperature range from −86 to +100 °C
  • Autoclavable when open (121 °C, 20 min.) for reliable sterilization
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity

Eppendorf® Safe-Lock 1.5 ml microtubes exemplify secure sample storage with their patented Safe-Lock design, preventing accidental openings and preserving sample integrity. Versatile across laboratory applications, the durable construction and 1.5 ml capacity cater to varied sample volumes. Clear graduations and a smooth surface enhance user-friendliness, ensuring a seamless and reliable laboratory experience. Elevate scientific endeavors with the trusted quality of Eppendorf® Safe-Lock tubes.

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Cap type: Safe-Lock
Material: PP
Volume: 1.5 ml

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.

Documents

Spécifications
  • Color:
    Blue
  • Packaging:
    500 tubes (1 bag×500 tubes)
  • Maximum RCF:
    30000×g
  • Cat. No.:
    CA21008-953
  • Temperature range:
    −86…+100 °C
  • Autoclavable:
    When open 121 °C, 20 mins