Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eppendorf® Flex-Tubes® Microtubes, 1.5 ml | Avantor

Eppendorf® Flex-Tubes® Microtubes, 1.5 ml

Blue, 500 tubes (1 bag × 500 tubes)
Catalog # CA20901-424
Supplier:   Eppendorf
Fournisseur #:  022363531
Eppendorf® Flex-Tubes® Microtubes, 1.5 ml
Catalog # CA20901-424
Supplier:  Eppendorf
Eppendorf® Flex-Tubes® Microtubes, 1.5 ml
Eppendorf® Flex-Tubes® Microtubes, 1.5 ml

Blue, 500 tubes (1 bag × 500 tubes)

Price
$113.00
Log in pour la disponibilité
Case of 500
Product Details & Documents

Flex-Tube® microtubes (1.5 ml) are designed for seamless one-handed operation with an easy-to-open, securely closing tube design. Perfect for sample preparation, centrifugation, and storage, these tubes will effortlessly integrate into workflows for efficient and reliable performance.

  • Easy-to-use ergonomic design featuring a user-friendly tube lid
  • Durable polypropylene construction capable of withstanding chemicals, mechanical stress, and extreme temperatures
  • g-Safe® technology ensures stability up to 30000×g, preventing sample loss during centrifugation and enhancing safety with hazardous samples
  • Free from slip agents, plasticizers, and biocides; eliminating substances that may compromise bio-assay results
  • Clear graduation marks allow for visual volume verification during pipetting
  • Convenient frosted lid and side surface facilitate easy labeling
  • Functional across a wide temperature range (−86 to 100 °C) for versatile applications
  • Autoclavable up to 121 °C for 20 minutes, ensuring effective sterilization
  • Available in both Eppendorf Quality and PCR Clean purity grades with lot certification

The easy-to-open, securely closing, Flex-Tube® offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry.

Eppendorf consumables, including the Flex-Tube®, are made exclusively from high-quality, ultra-pure polypropylene, which complies with FDA guidelines 21 CFR § 177.1520 »Olefin Polymers«, and 21 CFR § 178.2010 »Antioxidants and Stabilizers for Polymers«. Extensive quality checks are in place prior to and during production. Furthermore, the final product is examined for functionality.

The functionality tests for tubes include lid closure security in a boiling water bath, centrifugation stability, and vapor tightness. These quality controls and tests ensure the sustained high product quality across the entire production phase - lot by lot.

Highest sample integrity.
Manufactured without the use of slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results or measurements, e.g. nucleic acid quantification by photometric measurements at 260 nm.

Cap type: Flex-Tube® lid
Material: PP

Certifications: Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001), Lot-specific certficates for the purity grade PCR clean can be downloaded from the Eppendorf website.

Spécifications
  • Color:
    Blue
  • Packaging:
    500 tubes (1 bag × 500 tubes)
  • Cat. No.:
    CA20901-424
  • Operating temperature range:
    −86...100 °C
  • Volume:
    1.5 ml
  • Max. RCF:
    30000×g
  • Autoclavable:
    When open 121 °C, 20 min