Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipettes, Straight, 12 mL
Catalog # CA16007-002
Supplier: Electron Microscopy Science
Fournisseur #: 70986-00
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Catalog # CA16007-002
Supplier: Electron Microscopy Science
Jump to:
Product Details & Documents
Transfer pipette that works as a syringe for liquid transfer. Each pipette has calibration markings from 0 to 12 ml in 0.2 ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.
- Inexpensive and disposable
Spécifications
- Désignation:Transfer Pipettes, Straight, 12 mL