$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EC3 DIGITAL CAMERA W/SOFTWARE
Catalog # CA14400-000
Supplier:
Leica Microsystems
Expand image
EC3 DIGITAL CAMERA W/SOFTWARE
Catalog # CA14400-000
Supplier:
Leica Microsystems
Supplier Number:
12730079
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International