Sampling Tubes and Systems, Drager
Activated charcoal tubes, Type NIOSH
Catalog # CA13453-470
Supplier: Draeger
Fournisseur #: 6728631
Jump to:
Product Details & Documents
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
Spécifications
- Désignation:Activated charcoal tubes, Type NIOSH
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.