Sampling Tubes and Systems, Drager
Activated charcoal tubes, Type B/G
CA13453-468
: Draeger
: 8101821
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.
- Désignation:Activated charcoal tubes, Type B/G