Simultaneous Test Sets, Drager Safety
Simultaneous test set I, for inorganic fumes (acid gas, hydrocyanic acid, carbon monoxide, basic gas, nitrous gas), 40 s
Catalog # CA13453-124
Supplier: Draeger
Fournisseur #: 8101735
Jump to:
Product Details & Documents
Risks and potential dangers at a glance: The SimultanTest Sets allow the measurement of five different gases at the same time.
In the SimultanTest Set, five Dräger-Tubes are arranged next to each other in a stable rubber sleeve. The air to be monitored is simultaneously drawn in through the tubes by a Dräger-Tube pump. Quick determinations are made about the presence of dangerous substances at TLV levels.
The device delivers fast and reliable results for different combinations of organic and inorganic gases.
Spécifications
- Désignation:Simultaneous test set I, for inorganic fumes (acid gas, hydrocyanic acid, carbon monoxide, basic gas, nitrous gas)
- Response Time:40 s
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Extension Hose
Informations sur le produit
- Catalog #AvailabilityPrice
- Adapter for Simultaneous test sets, for connection to a bellows pumpAdapter for Simultaneous test sets, for connection to a bellows pumpCA34330-044Log in pour la disponibilitéEach - $595.27