Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
MetaPhor® Agarose

MetaPhor® Agarose

MetaPhor® agarose, 125 g, 2% DNA resolution between 200 and 800 bp; ≤600 kDa, 18...26 °C (64...79 °F)
Catalog # CA12001-916
Supplier:   Lonza
Supplier #:  50180
MetaPhor® Agarose
Catalog # CA12001-916
Supplier:  Lonza
CAS Number:  
MetaPhor® Agarose
MetaPhor® Agarose

MetaPhor® agarose, 125 g, 2% DNA resolution between 200 and 800 bp; ≤600 kDa, 18...26 °C (64...79 °F)

CAS Number:  
Price
$1,234.52
Log in pour la disponibilité
Each (125g)
Minimum order quantity is 1
Product Details & Documents

MetaPhor® is an intermediate melting temperature agarose that lets you resolve DNA fragments differing in size by 2%, in the range of 200 bp to 800 bp.

  • All Lonza agarose is quality tested to certify performance
  • High gel strength
  • Standard melting temperature

MetaPhor® Agarose is a high resolution agarose that challenges polyacrylamide. MetaPhor® Agarose is an intermediate melting temperature (75 °C) agarose with twice the resolution capabilities of the finest-sieving agarose products. For example, a 200 bp DNA fragment can be separated from a 204 bp fragment. Using fast running protocols DNA differing in size by 1% can be resolved in as little as 1.5 hours in a 20 cm long horizontal or vertical gel format. MetaPhor agarose gels (2 to 4%) approximate the resolution of polyacrylamide gels (4 to 8%). These agarose gels are ideal for resolving AMPFLPs, STRs, and tri- and tetranucleotide repeats. Ideal for small PCR fragments, MetaPhore® is the choice agarose for resolving hard to see bands.

Applications: Small fragments, PCR.

Spécifications

Spécifications

  • Désignation:
    MetaPhor® agarose
  • Taille:
    125 g
  • Température de stockage:
    18...26 °C (64...79 °F)
  • Utilisation:
    2% DNA resolution between 200 and 800 bp; ≤600 kDa

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.