Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
AccuGENE™ Buffers

AccuGENE™ Buffers

SSPE buffer (20X), 1 L, 7.4
Catalog # CA12001-394
Supplier:   Lonza
Supplier #:  51214
AccuGENE™ Buffers
Catalog # CA12001-394
Supplier:  Lonza
CAS Number:  
AccuGENE™ Buffers
AccuGENE™ Buffers

SSPE buffer (20X), 1 L, 7.4

CAS Number:  
Price
$159.81
Log in pour la disponibilité
Each
Product Details & Documents

AccuGENE® buffers are prepared with high quality reagents and use 18 megohm water

  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
  • Formulation - 3.0 M NaCl, 0.3 M sodium citrate, pH 7.0

Store all reagents at 18 to 26 °C (64 to 79 °F).

They are manufactured according to strict quality control standards to ensure lot-to-lot consistency.

Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.

Spécifications

Spécifications

  • Désignation:
    SSPE buffer (20X)
  • Taille:
    1 L
  • pH:
    7.4
  • Cat. No.:
    CA12001-394
  • Formulation:
    20X SSPE with 3.0 M NaCl, 0.2 M NaH2PO4 ò H2O, and 0.02 M EDTA

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.