Technical Buffer Solutions, Sachets
NIST-traceable buffer for pH electrode calibration and verification.
- NIST-traceable
- Ensure freshness
- Improve data
Also available in multiple packaging options.
Mettler Toledo buffer sachets ensure a fresh portion of buffer is used for each calibration, improving your pH data.
- Type de conditionnement:Sachet, green
- Taille:30×20 ml
- pH:7.00
- pH Accuracy:±0.02