Round Bottom Flasks
5 ml, 10/19, Round
Catalog # CA11007-822
 :  015890710
Round Bottom Flasks
Catalog # CA11007-822
Supplier:  
Round Bottom Flasks
5 ml, 10/19, Round
Discontinued
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
  • ST Joint No.:
    10/19
  • Capacité:
    5 ml
  • Bottom Style:
    Round