Round Bottom Flasks
Round Bottom Flasks
10 ml, 10/19, Round
  CA11007-818
Supplier #:  015890690
Round Bottom Flasks
  CA11007-818
 :  
Round Bottom Flasks
10 ml, 10/19, Round
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
  • ST Joint No.:
    10/19
  • Capacité:
    10 ml
  • Bottom Style:
    Round