Round Bottom Flasks
Round Bottom Flasks
25 ml, 19/26, Round
Catalog # CA11007-814
Supplier #:  015890670
Round Bottom Flasks
Catalog # CA11007-814
Supplier:  
Round Bottom Flasks
25 ml, 19/26, Round
Discontinued
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
  • ST Joint No.:
    19/26
  • Capacité:
    25 ml
  • Bottom Style:
    Round