Round Bottom Flasks
150 ml, 19/26, Round
Catalog # CA11007-802
 :  015890610
Round Bottom Flasks
Catalog # CA11007-802
Supplier:  
Round Bottom Flasks
150 ml, 19/26, Round
Discontinued
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
  • ST Joint No.:
    19/26
  • Capacité:
    150 ml
  • Bottom Style:
    Round