Polyethylene Plug Caps, Starplex® | Avantor
Polyethylene Plug Caps, Starplex®
Plug Style Caps, Green, Non sterile
Catalog # CA11007-698
Supplier:   Starplex
Fournisseur #:  V9200
Polyethylene Plug Caps, Starplex®
Catalog # CA11007-698
Supplier:  Starplex
Polyethylene Plug Caps, Starplex®
Plug Style Caps, Green, Non sterile
Price
$638.00
Log in pour la disponibilité
Case of 6,000
Product Details & Documents
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Spécifications
  • Sterility:
    Non sterile
  • Désignation:
    Plug Style Caps
  • Cap Color:
    Green