Polyethylene Plug Caps, Starplex®
Polyethylene Plug Caps, Starplex®
Plug Style Caps, Red, Non sterile
Catalog # CA11007-696
Supplier:   Starplex
Supplier #:  V9195
Polyethylene Plug Caps, Starplex®
Catalog # CA11007-696
Supplier:  Starplex
Polyethylene Plug Caps, Starplex®
Plug Style Caps, Red, Non sterile
Price
$638.00
Log in for availability
Case of 6,000
Product Details & Documents
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Specifications
  • Sterility:
    Non sterile
  • Désignation:
    Plug Style Caps
  • Cap Color:
    Red