qScript™ cDNA Synthesis Kits, QuantaBio
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The qScript cDNA Synthesis Kit is a sensitive and easy-to-use solution for RNA quantification using two step RT-qPCR.
- Economical 2-component kit ideally suited for high throughput expression studies
- Optimized, double pre-primed master mix component ensures equal representation of 5′ and 3′ RNA sequences
- Broad linear dynamic detection range with limiting (10pg) or plentiful (1ug) samples of total RNA
The qScript™ reaction mix provides all the necessary components for cDNA synthesis except the enzyme and RNA template. The optimized blend of random and oligo(dT) primers provides robust, consistent, and unbiased first-strand synthesis over a broad range of RNA template concentrations. qScript™ reverse transcriptase is a mixture of an engineered MMLV-RT and a ribonuclease inhibitor protein. The simplified reaction procedure is ideally suited for high throughput expression studies using RT-PCR. The resulting cDNA product is directly compatible with current real-time PCR methods or conventional end-point RT-PCR where the targets are ≤1kb in length.
Spécifications
Spécifications
- Désignation:qScript™ cDNA Synthesis Kits
- No. of Reactions:500