Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Tris-HCl Buffer, 1.5 M Sterile Solution

Tris-HCl Buffer, 1.5 M Sterile Solution

Tris-HCl Buffer 1.5 M, 1 L, 8.8
Catalog # CA101320-670
Supplier:   Teknova
Supplier #:  T1588
Tris-HCl Buffer, 1.5 M Sterile Solution
Catalog # CA101320-670
Supplier:  Teknova
CAS Number:  
Tris-HCl Buffer, 1.5 M Sterile Solution

Tris-HCl Buffer 1.5 M, 1 L, 8.8

CAS Number:  
Price
$363.63
Log in for availability
Each (1l)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

1.5 M Tris (hydroxymethyl) Aminomethane solution adjusted to pH 8.8.

Specifications

Specifications

  • Désignation:
    Tris-HCl Buffer 1.5 M
  • Taille:
    1 L
  • pH:
    8.8
  • Cat. No.:
    CA101320-670
  • Storage temperature:
    Store at room temperature. Shipped at ambient temperature.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.