Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, 0.2M, Specify, 500 ml
  CA100504-840
 :  11652
Sodium Cacodylate Buffer, Electron Microscopy Sciences
  CA100504-840
 :  
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, 0.2M, Specify, 500 ml
 :  
$1,502.44
Each (500ml)
Used for preparing buffered glutaraldehdye fixatives for electron microscopy
Available with a pH of 7.2 or 7.4.
Sodium Cacodylate Buffer, 0.2M is prepared from Sodium Cacodylate Trihydrate (F.W 214.02).
  • Désignation:
    Sodium Cacodylate Buffer, 0.2M
  • Taille:
    500 ml
  • pH:
    Specify