Sodium Cacodylate Buffer, Electron Microscopy Sciences | Avantor
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, 0.2M, Specify, 225 ml
Catalog # CA100503-718
Fournisseur #:  11650
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Catalog # CA100503-718
CAS Number:  
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, 0.2M, Specify, 225 ml
CAS Number:  
Price
$702.54
Log in pour la disponibilité
Each (225ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Used for preparing buffered glutaraldehdye fixatives for electron microscopy
Available with a pH of 7.2 or 7.4.
Sodium Cacodylate Buffer, 0.2M is prepared from Sodium Cacodylate Trihydrate (F.W 214.02).
Spécifications
Spécifications
  • Désignation:
    Sodium Cacodylate Buffer, 0.2M
  • Taille:
    225 ml
  • pH:
    Specify