YPD Plates
YPD Plates with 1.2 M Sorbitol, 100 mm
Catalog # CA100219-702
Supplier: Teknova
Supplier #: Y1010
Jump to:
Product Details & Documents
YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.
- 20 Plates
- Sterile
Ingredients: 1.0% Yeast Extract (10.0 g/L), 2.0% Peptone (20.0 g/L), 2.0% Glucose (20.0g/L), 2.0% Agar (20.0g/L).
Ambient shipping conditions. Store at room temperature or 2–8°C.
Specifications
Specifications
- Désignation:YPD Plates with 1.2 M Sorbitol
- Taille:100 mm
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



