Tryptic Soy Agar Plates
Tryptic soy agar plates, 86×128 mm, 20 plates
Catalog # CA100218-562
Supplier: Teknova
Supplier #: T0147
Jump to:
Product Details & Documents
Tryptic Soy Agar Plates contains 1.5% Tryptone, 0.5% soytone, 0.5% NaCl, and 1.5% agar used for isolating and cultivating a variety of fastidious microorganisms.
Specifications
Specifications
- Désignation:Tryptic soy agar plates
- Taille:20 plates
- Plate Size:86×128 mm
- Cat. No.:CA100218-562
- Sterility:Sterile



