Galactose Solution
Galactose Solution, 20%, 100 mL
CA100216-726
: G0500
For used in growth media and stored at room temperature.
- Désignation:Galactose Solution, 20%
- Taille:100 mL
- Cat. No.:CA100216-726
- Sterility:Sterile
For used in growth media and stored at room temperature.