YPD Plates
YPD Agar Plates, 40 mL/Plates, 100 mm
Catalog # CA100216-448
Supplier: Teknova
: Y1030
Jump to:
Product Details & Documents
YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.
- 20 Plates
- Sterile
Ingredients: 1.0% Yeast Extract (10.0 g/L), 2.0% Peptone (20.0 g/L), 2.0% Glucose (20.0g/L), 2.0% Agar (20.0g/L).
Ambient shipping conditions. Store at room temperature or 2–8°C.
Spécifications
Spécifications
- Désignation:YPD Agar Plates, 40 mL/Plates
- Taille:100 mm
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



