Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
YPD Plates | Avantor

YPD Plates

YPD Agar, 100 mm
Catalog # CA100216-446
Supplier:   Teknova
Fournisseur #:  Y1000
YPD Plates
Catalog # CA100216-446
Supplier:  Teknova
CAS Number:  
YPD Plates
YPD Plates

YPD Agar, 100 mm

CAS Number:  
Price
$556.99
Log in pour la disponibilité
Pack of 20
La quantité minimale de commande est de 1
Product Details & Documents

YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.

  • 20 Plates
  • Sterile

Ingredients: 1.0% Yeast Extract (10.0 g/L), 2.0% Peptone (20.0 g/L), 2.0% Glucose (20.0g/L), 2.0% Agar (20.0g/L).

Ambient shipping conditions. Store at room temperature or 2–8°C.

Spécifications

Spécifications

  • Désignation:
    YPD Agar
  • Taille:
    100 mm

Spécifications