BD BBL™ Isolator Pack Plated Media, BD Diagnostics
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
BD BBL™ prepared agar plates in isolator packs are plated media in gamma-irradiated packs that are specially designed for isolator systems
- Specially designed for isolator systems
- Validated for performance and sterility according to the Association for the Advancement of Medical Instrumentation (AAMI) guidelines
To use, place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle. When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping. Use the included inner bag to transport plates from your critical environment. Storage temperature: 2 to 8 °C.
The multi-wrap packaging protects media from vaporized hydrogen peroxide exposure.
Ordering information: Each pack or case contains a signed, lot-specific Certificate of Analysis.
Spécifications
Spécifications
- Type de conditionnement:Isolator pack, triple wrapping, VHP resistant
- Désignation:Trypticase™ Soy Agar with Lecithin and Polysorbate 80, Finger Dab
- Plate Diameter:100 mm
- Cat. No.:CA-90007-914
- Sterility:Sterile
Spécifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



