Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
N-Boc-L-threonine

N-Boc-L-threonine

100 g
  A-2305.0100BA
Supplier #:  4000124.0100
N-Boc-L-threonine
  A-2305.0100BA
 :  2592-18-9
N-Boc-L-threonine

100 g

 :  2592-18-9
 
$507.68
   
Each (100g)
Minimum order quantity is 1
 

Sequence: Boc-Thr-OH

Bachem also offers phosphothreonine derivatives.

Building block for the incorporation of O-acyl isopeptide bonds during Fmoc-SPPS.

 :  N-Boc-L-threonine, Boc-Thr-OH, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid
  •  :2592-18-9
  •  :C₉H₁₇NO₅
  •  :MFCD00065946
  •  :219.2 g/mol
  •  :Réfrigérateur
  •  :219-987-6
 

 

  • Taille:
    100 g

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.