Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
N-Boc-L-threonine

N-Boc-L-threonine

25 g
Catalog # A-2305.0025BA
Supplier:   Bachem Americas
Supplier #:  4000124.0025
N-Boc-L-threonine
Catalog # A-2305.0025BA
Supplier:  Bachem Americas
CAS Number:  2592-18-9
N-Boc-L-threonine

25 g

CAS Number:  2592-18-9
Price
$194.26
Log in for availability
Each (25g)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequence: Boc-Thr-OH

Bachem also offers phosphothreonine derivatives.

Building block for the incorporation of O-acyl isopeptide bonds during Fmoc-SPPS.

Synonyms:  N-Boc-L-threonine, Boc-Thr-OH, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid
  • CAS Number:2592-18-9
  • Formula:C₉H₁₇NO₅
  • MDL Number:MFCD00065946
  • Molecular Weight:219.2 g/mol
  • Storage Temperature:Réfrigérateur
  • EINECS:219-987-6
Specifications

Specifications

  • Taille:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.