Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, 8 mm Hole, Aluminum, White Silicone/ Aluminum Foil, 50° Shore A, 1.3 mm, Silver
Catalog # 97057-728
Supplier: MicroSolv Technology Corp
Fournisseur #: 95020-25S-T
Jump to:
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Spécifications
- Septum:White Silicone/ Aluminum Foil, 50° Shore A, 1.3 mm
- Color:Silver
- Désignation:Magnetic Screw Cap
- Matière:Aluminum
- Taille:18 mm Precision Threads, 8 mm Hole